MilliporeSigma
  • Structural basis of recognition of interferon-α receptor by tyrosine kinase 2.
Structural basis of recognition of interferon-α receptor by tyrosine kinase 2.
Nature structural & molecular biology (2014-04-08)
Heidi J A Wallweber, Christine Tam, Yvonne Franke, Melissa A Starovasnik, Patrick J Lupardus
ABSTRACT
Tyrosine kinase 2 (TYK2) is a member of the Janus kinase (JAK) family of nonreceptor tyrosine kinases, which are essential for proper signaling in immune responses and development. Here we present a 2.0-Å-resolution crystal structure of a receptor-binding fragment of human TYK2, encompassing the FERM and SH2 domains, in complex with a so-called 'box2'-containing intracellular peptide motif from the interferon-α receptor chain 1 (IFNAR1). The TYK2-IFNAR1 interface reveals an unexpected receptor-binding mode that mimics a SH2 domain-phosphopeptide interaction, with a glutamate replacing the canonical phosphotyrosine residue. This structure provides the first view, to our knowledge, of a JAK in complex with its cognate receptor and defines the molecular logic through which JAKs have evolved to interact with divergent receptor sequences.