MilliporeSigma
  • Mitsugumin 56 (hedgehog acyltransferase-like) is a sarcoplasmic reticulum-resident protein essential for postnatal muscle maturation.
Mitsugumin 56 (hedgehog acyltransferase-like) is a sarcoplasmic reticulum-resident protein essential for postnatal muscle maturation.
FEBS letters (2015-04-07)
Bo Van, Miyuki Nishi, Shinji Komazaki, Atsuhiko Ichimura, Sho Kakizawa, Keita Nakanaga, Junken Aoki, Ki-Ho Park, Jianjie Ma, Tomomi Ueyama, Takehiro Ogata, Naoki Maruyama, Hiroshi Takeshima
ABSTRACT
Mitsugumin 56 (MG56), also known as the membrane-bound O-acyl-transferase family member hedgehog acyltransferase-like, was identified as a new sarcoplasmic reticulum component in striated muscle. Mg56-knockout mice grew normally for a week after birth, but shortly thereafter exhibited a suckling defect and died under starvation conditions. In the knockout skeletal muscle, regular contractile features were largely preserved, but sarcoplasmic reticulum elements swelled and further developed enormous vacuoles. In parallel, the unfolded protein response was severely activated in the knockout muscle, and presumably disrupted muscle development leading to the suckling failure. Therefore, MG56 seems essential for postnatal skeletal muscle maturation.
Sigma-Aldrich
Monoclonal Anti-Dihydropyridine Receptor (α1 Subunit) antibody produced in mouse, clone 1A, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution