MilliporeSigma
Injectable controlled release depots for large molecules.
Journal of controlled release : official journal of the Controlled Release Society (2014-06-15)
Steven P Schwendeman, Ronak B Shah, Brittany A Bailey, Anna S Schwendeman
ABSTRACT
Biodegradable, injectable depot formulations for long-term controlled drug release have improved therapy for a number of drug molecules and led to over a dozen highly successful pharmaceutical products. Until now, success has been limited to several small molecules and peptides, although remarkable improvements have been accomplished in some of these cases. For example, twice-a-year depot injections with leuprolide are available compared to the once-a-day injection of the solution dosage form. Injectable depots are typically prepared by encapsulation of the drug in poly(lactic-co-glycolic acid) (PLGA), a polymer that is used in children every day as a resorbable suture material, and therefore, highly biocompatible. PLGAs remain today as one of the few "real world" biodegradable synthetic biomaterials used in US FDA-approved parenteral long-acting-release (LAR) products. Despite their success, there remain critical barriers to the more widespread use of PLGA LARproducts, particularly for delivery of more peptides and other large molecular drugs, namely proteins. In this review, we describe key concepts in the development of injectable PLGA controlled-release depots for peptides and proteins, and then use this information to identify key issues impeding greater widespread use of PLGA depots for this class of drugs. Finally, we examine important approaches, particularly those developed in our research laboratory, toward overcoming these barriers to advance commercial LAR development.
Sigma-Aldrich
Goserelin acetate, ≥99% (HPLC), white, powder
Goserelin, European Pharmacopoeia (EP) Reference Standard
Goserelin for NMR identification, European Pharmacopoeia (EP) Reference Standard