MilliporeSigma
  • MYC impairs resolution of site-specific DNA double-strand breaks repair.
MYC impairs resolution of site-specific DNA double-strand breaks repair.
Mutation research (2015-03-17)
Susanna Ambrosio, Stefano Amente, Giuliana Napolitano, Giacomo Di Palo, Luigi Lania, Barbara Majello
ABSTRACT
Although it is established that when overexpressed, the MYC family proteins can cause DNA double-stand breaks (DSBs) and genome instability, the mechanisms involved remain unclear. MYC induced genetic instability may result from increased DNA damage and/or reduced DNA repair. Here we show that when overexpressed, MYC proteins induce a sustained DNA damage response (DDR) and reduce the wave of DSBs repair. We used a cell-based DSBs system whereby, upon induction of an inducible restriction enzyme AsiSI, hundreds of site-specific DSBs are generated across the genome to investigate the role of MYC proteins on DSB. We found that high levels of MYC do not block accumulation of γH2AX at AsiSI sites, but delay its clearance, indicating an inefficient repair, while the initial recognition of DNA damage is largely unaffected. Repair of both homologous and nonhomologous repair-prone segments, characterized by high or low levels of recruited RAD51, respectively, was delayed. Collectively, these data indicate that high levels of MYC proteins delay the resolution of DNA lesions engineered to occur in cell cultures.
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture