MilliporeSigma
  • Synthesis of atactic and isotactic poly(1,2-glycerol carbonate)s: degradable polymers for biomedical and pharmaceutical applications.
Synthesis of atactic and isotactic poly(1,2-glycerol carbonate)s: degradable polymers for biomedical and pharmaceutical applications.
Journal of the American Chemical Society (2013-04-25)
Heng Zhang, Mark W Grinstaff
ABSTRACT
The synthesis and characterization of atactic and isotactic linear poly(benzyl 1,2-glycerol carbonate)s are reported. The poly(benzyl 1,2-glycerol carbonate)s were obtained via the ring-opening copolymerization of rac-/(R)-benzyl glycidyl ether with CO2 using [SalcyCo(III)X] complexes with high carbonate linkage selectivity and polymer/cyclic carbonate selectivity (>99%). Deprotection of the resultant polymers afforded poly(1,2-glycerol carbonate)s with a functionalizable pendant primary hydroxyl group. Poly(1,2-glycerol carbonate) showed a remarkable increase in degradation rate compared to poly(1,3-glycerol carbonate) with a t1/2 ≈ 2-3 days. These polymers fulfill an unmet need for a readily degradable biocompatible polycarbonate.
Sigma-Aldrich
Benzyl glycidyl ether, 99%