MilliporeSigma
  • Enantioseparation of 1-arylethanols via a supramolecular chiral host consisting of N-(2-naphthoyl)-L-aspartic acid and an achiral diamine.
Enantioseparation of 1-arylethanols via a supramolecular chiral host consisting of N-(2-naphthoyl)-L-aspartic acid and an achiral diamine.
Organic & biomolecular chemistry (2012-01-26)
Koichi Kodama, Ayaka Kanno, Eriko Sekine, Takuji Hirose
ABSTRACT
A supramolecular chiral host consisting of N-(2-naphthoyl)-L-aspartic acid (L-1) and meso-1,2-diphenylethylenediamine (2) is effective in enantioseparation of 1-arylethanols (up to 96% ee with 100% inclusion ratio). Here we report three different methods to prepare the inclusion crystals and discuss the chiral recognition mechanism on the basis of X-ray crystallography results.
Sigma-Aldrich
meso-1,2-Diphenylethylenediamine, 98%