MilliporeSigma
  • 4-Thiazolidinones in heterocyclic synthesis: synthesis of novel enaminones, azolopyrimidines and 2-arylimino-5-arylidene-4-thiazolidinones.
4-Thiazolidinones in heterocyclic synthesis: synthesis of novel enaminones, azolopyrimidines and 2-arylimino-5-arylidene-4-thiazolidinones.
Molecules (Basel, Switzerland) (2012-05-29)
Haider Behbehani, Hamada Mohamed Ibrahim
ABSTRACT
The 4-thiazolidinones 3a-d were used as a key intermediates for the synthesis of 2-arylimino-5-arylidene-4-thiazolidinones derivatives 7a–p via nucleophilic addition reactions with the arylidene malononitrile. Moreover the 4-thiazolidinones 3a and 3c condensed with the DMF-DMA to form the corresponding enamines 8 and 9 depending on the reaction conditions. Otherwise the 4-thiazolidinone 3b reacts regioselectively with DMF-DMA to afford the enaminones 10 and 11, respectively. The latter reacts with many heterocyclic amines affording polyfunctionally substituted fused pyrimidine derivatives 13-18. The enamine 8b was also reacted with the 3-amino-1,2,4-triazole to afford the acyclic product 19, which could not be further cyclized to the corresponding tricyclic system 20. Moreover the 4-thiazolidinone 3c reacted with the benzenediazonium chloride to afford the arylhydrazones 12. The X-ray single crystal technique was employed in this study for structure elucidation and Z/E potential isomerism configuration determination. The X-ray crystallographic analyses of eight products could be obtained, thus establishing with certainty the structures proposed in this work.