MilliporeSigma
  • Monitoring of cellulose depolymerization in 1-ethyl-3-methylimidazolium acetate by shear and elongational rheology.
Monitoring of cellulose depolymerization in 1-ethyl-3-methylimidazolium acetate by shear and elongational rheology.
Carbohydrate polymers (2014-12-17)
Anne Michud, Michael Hummel, Simon Haward, Herbert Sixta
ABSTRACT
The thermal stability of cellulose in the ionic liquid (IL) 1-ethyl-3-methylimidazolium acetate, [emim]OAc was investigated. For this purpose, Eucalyptus urugrandis prehydrolysis kraft pulp was first dissolved in [emim]OAc by means of a vertical kneader and then stored at three different temperatures to study the time-depended behavior of the cellulose-[emim]OAc system. Cellulose depolymerization was assessed by characterizing the precipitated cellulose and the rheological behavior of the cellulose-[emim]OAc solutions. The results show decreases in the weight average molecular mass and in the shear viscosity at temperatures exceeding 60 °C, which can be related to progressing degradation of cellulose in the IL upon storage at elevated temperature. The changes in behavior of the solutions under extensional stresses also attest the gradual depolymerization of cellulose. The degradation has been analyzed using appropriate kinetic models. Propyl gallate appeared to be an efficient stabilizer of the cellulose-[emim]OAc system during the dissolution step even though the mechanism has not been fully understood yet.
Sigma-Aldrich
1-Ethyl-3-methylimidazolium acetate, ≥95.0% (HPLC)