MilliporeSigma
  • Pharmaceutical-enantiomers resolution using immobilized polysaccharide-based chiral stationary phases in supercritical fluid chromatography.
Pharmaceutical-enantiomers resolution using immobilized polysaccharide-based chiral stationary phases in supercritical fluid chromatography.
Journal of chromatography. A (2014-01-21)
Katrijn De Klerck, Yvan Vander Heyden, Debby Mangelings
ABSTRACT
Since their introduction on the market the applicability of immobilized polysaccharide-based chiral stationary phases in high-performance liquid chromatography has been thoroughly investigated. These immobilized phases have the benefit to be applicable with a wide range of modifiers, potentially extending the application range of the polysaccharide-based stationary phases. Because an increasing number of stationary phases are being introduced in the field of chiral chromatography it is important to evaluate their enantioselectivity in different techniques in order to get an idea about their applicability. In this study, three immobilized chiral polysaccharide-based stationary phases (Chiralpak IA, IB, and IC) are evaluated in supercritical fluid chromatography (SFC) with a test set of pharmaceutical racemates. This is done in a three-fold manner: their performance is evaluated (1) using traditional modifiers, (2) using mixtures of atypical modifiers, and (3) the results were compared to those on coated stationary phases with an equivalent chiral selector. To get a visual overview of the enantioselective patterns of the different chromatographic systems (mobile and stationary phase combinations), a Principal Component Analysis is performed, which allows determining the (dis)similarity between individual systems. To assess the complementarity cumulative success rates are determined. The immobilized chiral stationary phases prove to yield high cumulative success rates.
Product Description
Supelco
Astec® Cellulose DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® Cellulose DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® Cellulose DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm