MilliporeSigma
Environmental impact of diuron transformation: a review.
Chemosphere (2004-07-28)
S Giacomazzi, N Cochet
ABSTRACT
Diuron is a biologically active pollutant present in soil, water and sediments. A synthesis of literature data on its physicochemical properties, partitioning behaviour, abiotic and biotic transformations, toxicological and ecotoxicological impacts has been here performed. Data have shown that diuron is generally persistent in soil, water and groundwater. It is also slightly toxic to mammals and birds as well as moderately toxic to aquatic invertebrates. However, its principal product of biodegradation, 3,4-dichloroaniline exhibits a higher toxicity and is also persistent in soil, water and groundwater. Thus, diuron indirectly possesses a significant amount of toxicity and could be a potential poisoning pesticide contaminant of groundwater. Unfortunately, groundwater contamination will still persist despite the progressive suppression of diuron (Directive 200/60/CE). Therefore, determining the main factors influencing its degradation and its ecotoxicological effects on the environment and health could provide a basis for further development of bioremediation processes.
Product Description
Supelco
Diuron, PESTANAL®, analytical standard
Sigma-Aldrich
Diuron, ≥98%