MilliporeSigma
  • Direct RNA sequencing mediated identification of mRNA localized in protrusions of human MDA-MB-231 metastatic breast cancer cells.
Direct RNA sequencing mediated identification of mRNA localized in protrusions of human MDA-MB-231 metastatic breast cancer cells.
Journal of molecular signaling (2013-09-06)
Kristine Raaby Jakobsen, Emilie Sørensen, Karin Kathrine Brøndum, Tina Fuglsang Daugaard, Rune Thomsen, Anders Lade Nielsen
ABSTRACT
Protrusions of cancer cells conferrers a vital function for cell migration and metastasis. Protein and RNA localization mechanisms have been extensively examined and shown to play pivotal roles for the functional presence of specific protein components in cancer cell protrusions. To describe genome wide RNA localized in protrusions of the metastatic human breast cancer cell line MDA-MB-231 we used Boyden chamber based methodology followed by direct mRNA sequencing. In the hereby identified group of protrusion localized mRNA some previously were described to be localized exemplified by mRNA for Ras-Related protein 13 (RAB13) and p0071 (Plakophilin-4/PKP4). For other transcripts, exemplified by mRNA for SH3PXD2A/TKS5 and PPFIA1/Liprin-α1, only the corresponding proteins previously were described to have protrusion localization. Finally, a cohort of MDA-MB-231 protrusion localized transcripts represents novel candidates to mediate cancer cell subcellular region specific functions through mRNA direction to protrusions. We included a further characterization of p0071, an armadillo repeat protein of adherence junctions and desmosomes, in MDA-MB-231 and non-metastatic MCF7 cells including analysis of novel identified alternative spliced p0071 mRNA isoforms. The results showed isoform and cell type specific p0071 mRNA localization. Altogether, the presented data represents a genome wide and gene specific descriptive and functional analyses of RNA localization in protrusions of MDA-MB-231 metastatic cancer cells.
Sigma-Aldrich
Anti-Actin, N-terminal antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution