MilliporeSigma
  • Relative potencies of the four stereoisomers of isomalathion for inhibition of hen brain acetylcholinesterase and neurotoxic esterase in vitro.
Relative potencies of the four stereoisomers of isomalathion for inhibition of hen brain acetylcholinesterase and neurotoxic esterase in vitro.
Toxicology and applied pharmacology (1996-08-01)
S Jianmongkol, C E Berkman, C M Thompson, R J Richardson
ABSTRACT
The cholinergic toxicity of malathion is exacerbated by its isomerization product, isomalathion, which inhibits detoxifying carboxylesterases as well as target acetylcholinesterase (AChE). Previous work has shown that the four stereoisomers of isomalathion, (1R, 3R), (1R, 3S), (1S, 3R), and (1S, 3S), differ in their inhibitory potencies against either rat brain or electric eel AChE. The present study examined the relative inhibitory potencies of these stereoisomers and the totally racemic mixture (1RS, 3RS) against hen brain AChE and neurotoxic esterase (NTE) to provide new data on stereoselective inhibition of neurotoxicologically significant esterases and to assess the potential of these compounds to cause organophosphorus (OP) compound-induced delayed neurotoxicity (OPIDN). The order of potencies against hen brain AChE was (1R, 3R) > (1R, 3S) > (1RS, 3RS) > (1S, 3R) > (1S, 3S), with a 15-fold difference between the strongest (ki = 388 mM-1 min-1; 20 min I50 = 89.3 nM) and weakest (ki = 25.6 mM-1 min-1; 20 min I50 = 1354 nM) inhibitors. Both asymmetric centers contributed substantially and interdependently to inhibitory potency, but the effect of changing the configuration at phosphorus alone was greater than changing the configuration at carbon alone. None of the isomalathions was an effective inhibitor of hen brain NTE (extrapolated 20 min I50 values were 1.2 to 29 mM), yielding NTE/ AChE I50 ratios (neuropathy target ratios, NTRs) of 1.5 x 10(3) to 1.5 x 10(5). NTRs of this magnitude indicate that none of the isomalathions should initiate OPIDN, even after doses greatly exceeding the LD50. Therefore, reports of OPIDN or other neuropathic sequelae associated with malathion exposures in humans cannot be explained on the basis of NTE inhibition by contaminating isomalathions.
Product Description
Malathion impurity A, European Pharmacopoeia (EP) Reference Standard