MilliporeSigma
  • Measurement of lymphatic flow variation by noninvasive method cases of lymphedema.
Measurement of lymphatic flow variation by noninvasive method cases of lymphedema.
Angiology (1992-07-01)
G Thibaut, A Durand, P Follignoni, A Bertrand
ABSTRACT
The purpose of this study was to measure the variations of lymphatic flow. A noninvasive isotopic method was used to achieve a functional exploration of lymphatic circulation. Fifteen subjects were used in the study: 10 healthy subjects and 5 patients with lower extremity lymphedema. A first subcutaneous injection of technetium 99m rhenium sulfate (99mTc) was performed in the first interdigital space of both feet. The radioactivity was recorded in two places: the first one on the inguinal site by a gamma camera; the second, below the first, on the precordial site by a multichannel analyzer. With the two types of recording procedures, it was possible to obtain a curve that showed the amount of radioactivity in relation to time. In order to obtain a muscular activity fifty-five minutes after the injection, each subject or patient spent ten minutes on an ergometric bike. A second subcutaneous injection was performed one week later, but prior to the injection, the subject or patient took orally 1800 mg of heptaminol adenosine phosphate (HAP) per day for three days. The radioactivity recording was made under the same conditions as with the muscular activity. The statistical results of the experiment without treatment on the two types of recording show that in the healthy subjects the amount of radioactivity increased during muscular activity. Moreover, the treatment indicated higher radioactivity values, which remained at a higher level. However, the muscular activity performed by a patient was unable to increase the radioactivity. On the other hand, the drug gave radioactivity values that were higher than the previous values of the first curve.(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
Heptaminol hydrochloride, European Pharmacopoeia (EP) Reference Standard