MilliporeSigma
  • In vivo induction of sister chromatid exchanges (SCEs) and chromosomal aberrations (CAs) by lynestrenol.
In vivo induction of sister chromatid exchanges (SCEs) and chromosomal aberrations (CAs) by lynestrenol.
Indian journal of experimental biology (2005-04-09)
Yasir Hasan Siddique, Mohammad Afzal
ABSTRACT
Genotoxicity study of synthetic progestin lynestrenol, was carried out on mouse bone marrow cells using sister chromatid exchanges (SCEs) and chromosomal aberrations (CAs) as parameters. Lynestrenol was studied at three different doses (6.87, 13.75 and 27.50 mg/kg body wt.). SCE and CA increased significantly as compared to normal control when treated with lynestrenol at 13.75 and 27.50 mg/kg body wt. The present results suggest that lynestrenol has both a genotoxic and cytotoxic effects in mouse bone marrow cells.
Product Description
Lynestrenol for peak identification, European Pharmacopoeia (EP) Reference Standard
Lynestrenol, European Pharmacopoeia (EP) Reference Standard