MilliporeSigma
  • Degradation of dibenzofuran via multiple dioxygenation by a newly isolated Agrobacterium sp. PH-08.
Degradation of dibenzofuran via multiple dioxygenation by a newly isolated Agrobacterium sp. PH-08.
Journal of applied microbiology (2013-11-28)
T T Le, K Murugesan, I-H Nam, J-R Jeon, Y-S Chang
ABSTRACT
To demonstrate the biodegradation of dibenzofuran (DF) and its structural analogs by a newly isolated Agrobacterium sp. PH-08. To assess the biodegradation potential of newly isolated Agrobacterium sp. PH-08, various substrates were evaluated as sole carbon sources in growth and biotransformation experiments. ESI LC-MS/MS analysis revealed the presence of angular degrading by-products as well as lateral dioxygenation metabolites in the upper pathway. The metabolites in the lower pathway also were detected. In addition, the cometabolically degraded daughter compounds of DF-related compounds such as BP and dibenzothiophene (DBT) in dual substrate degradation were observed. Strain PH-08 exhibited the evidence of meta-cleavage pathway as confirmed by the activity and gene expression of catechol-2,3-dioxygenase. Newly isolated bacterial strain, Agrobacterium sp. PH-08, grew well with and degraded DF via both angular and lateral dioxygenation as demonstrated by metabolites identified through ESI LC-MS/MS and GC-MS analyses. The other heterocyclic pollutants were also cometabolically degraded. Few reports have described the complete degradation of DF by a cometabolic lateral pathway. Our study demonstrates the novel results that the newly isolated strain utilized the DF as a sole carbon source and mineralized it via multiple dioxygenation.
Sigma-Aldrich
Dibenzofuran, 98%