MilliporeSigma
  • Au(I)/Ag(I)-catalyzed cascade approach for the synthesis of benzo[4,5]imidazo[1,2-c]pyrrolo[1,2-a]quinazolinones.
Au(I)/Ag(I)-catalyzed cascade approach for the synthesis of benzo[4,5]imidazo[1,2-c]pyrrolo[1,2-a]quinazolinones.
The Journal of organic chemistry (2013-04-06)
Xun Ji, Yu Zhou, Jinfang Wang, Linxiang Zhao, Hualiang Jiang, Hong Liu
ABSTRACT
An efficient and facile Au(I)/Ag(I)-catalyzed cascade method has been developed for one-pot synthesis of the complex polycyclic heterocycles benzo[4,5]imidazo[1,2-c]pyrrolo[1,2-a]quinazolinone derivatives through treatment of the substituted 2-(1H-benzo[d]imidazol-2-yl)anilines with 4-pentynoic acid or 5-hexynoic acid. The strategy features a Au(I)/Ag(I)-catalyzed one-pot cascade process involving the formation of three new C-N bonds in high yields, and with broad a substrate scope.
Sigma-Aldrich
4-Pentynoic acid, 95%