MilliporeSigma
  • A facile glovebox-free strategy to significantly accelerate the syntheses of well-defined polypeptides by
A facile glovebox-free strategy to significantly accelerate the syntheses of well-defined polypeptides by
Macromolecules (2013-06-25)
Jiong Zou, Jingwei Fan, Xun He, Shiyi Zhang, Hai Wang, Karen L Wooley
ABSTRACT
A facile N
Sigma-Aldrich
Hexylamine, 99%