MilliporeSigma
  • Microwave accelerated synthesis of isoxazole hydrazide inhibitors of the system xc- transporter: Initial homology model.
Microwave accelerated synthesis of isoxazole hydrazide inhibitors of the system xc- transporter: Initial homology model.
Bioorganic & medicinal chemistry letters (2013-09-18)
Afnan A Matti, Joseph Mirzaei, John Rudolph, Stephen A Smith, Jayme L Newell, Sarjubhai A Patel, Michael R Braden, Richard J Bridges, Nicholas R Natale
ABSTRACT
Microwave accelerated reaction system (MARS) technology provided a good method to obtain selective and open isoxazole ligands that bind to and inhibit the Sxc- antiporter. The MARS provided numerous advantages, including: shorter time, better yield and higher purity of the product. Of the newly synthesized series of isoxazoles the salicyl hydrazide 6 exhibited the highest level of inhibitory activity in the transport assay. A homology model has been developed to summarize the SAR results to date, and provide a working hypothesis for future studies.
Sigma-Aldrich
Isoxazole, 99%