MilliporeSigma
  • The metabolic activation of 4-nitro-o-phenylenediamine by chlorophyll-containing plant extracts: the relationship between mutagenicity and antimutagenicity.
The metabolic activation of 4-nitro-o-phenylenediamine by chlorophyll-containing plant extracts: the relationship between mutagenicity and antimutagenicity.
Mutation research (1991-09-01)
J M Gentile, G J Gentile
ABSTRACT
Chlorophyllin, the sodium and copper salt of chlorophyll, chlorophyll a, and chlorophyll b were tested for their ability to inhibit the mutagenic activity of the direct-acting mutagen 4-nitro-o-phenylenediamine (NOP) and its plant-activated mutagenic enhancement. All three forms of chlorophyll were antimutagenic against both NOP and its plant-activated product, with chlorophyllin proving most effective. Chlorophyll-containing plant extracts, however, proved very efficient at activating NOP into a mutagen of greater potency. When these extracts were assayed for total chlorophyll content it was found that they contained far less chlorophyll than was required for an antimutagenic effect to occur. Thus, the balance between chemical mutagen activation and/or enhancement by chlorophyll-containing plant extracts and the potential antimutagenicity of these plant extracts is a function of chlorophyll concentration. The data presented here indicate that this balance must be taken into consideration in future studies investigating the efficacy of complex natural plant extracts as antimutagenic substances.
Sigma-Aldrich
4-Nitro-o-phenylenediamine, 98%