MilliporeSigma
  • Organic acid excretion in a patient with 3-hydroxy-3-methylglutaryl-CoA lyase deficiency: facts and artefacts.
Organic acid excretion in a patient with 3-hydroxy-3-methylglutaryl-CoA lyase deficiency: facts and artefacts.
Clinica chimica acta; international journal of clinical chemistry (1978-12-01)
M Duran, D Ketting, S K Wadman, C Jakobs, R B Schutgens, H A Veder
PMID719902
ABSTRACT
The organic acid excretion in a patient with 3-hydroxy-3-methylglutaryl-CoA lyase deficiency is described. In addition to 3-hydroxy-3-methylglutaric, 3-methylglutaconic, 3-methylglutaric and 3-hydroxyisovaleric acids, substantial amounts of glutaric and adipic acids were repeatedly observed. 3-Methylcrotonic acid and possibly one of its isomers was found to be formed artificially from 3-methylglutaconic acid.
Sigma-Aldrich
3,3-Dimethylacrylic acid, 97%