MilliporeSigma
Fluorene-1-carboxylic acid.
Acta crystallographica. Section C, Crystal structure communications (1996-04-15)
A C Blackburn, A J Dobson, R E Gerkin
ABSTRACT
In fluorene-1-carboxylic acid, C14H10O2, the sole hydrogen bond is of the cyclic dimer type about a center of symmetry. The carboxyl H atom is ordered. Distances in the fluorene core are very similar to those in fluorene itself; the fluorene core dihedral angle is, however, larger than in fluorene.
Sigma-Aldrich
Fluorene-1-carboxylic acid, 97%