MilliporeSigma
  • Selective oxidation of 8,8'-hydroxylated binaphthols to bis-spironaphthalenones or binaphtho-para- and binaphtho-ortho-quinones.
Selective oxidation of 8,8'-hydroxylated binaphthols to bis-spironaphthalenones or binaphtho-para- and binaphtho-ortho-quinones.
Organic letters (2012-09-04)
Erin E Podlesny, Patrick J Carroll, Marisa C Kozlowski
ABSTRACT
The selective oxidation of a series of functionalized 8,8'-hydroxylated binaphthols to binaphtho-para- and binaphtho-ortho-quinones has been realized using either a Co-salen catalyst or ortho-iodoxybenzoic acid. A unique spirocyclic bis-spironaphthalenone was also obtained in good yield via a phenyliodonium diacetate promoted oxidative dearomatization.
Sigma-Aldrich
N,N′-Bis(salicylidene)ethylenediamine, 98%