MilliporeSigma
  • Hemodynamic and hormonal stress responses to endotracheal tube and ProSeal Laryngeal Mask Airway™ for laparoscopic gastric banding.
Hemodynamic and hormonal stress responses to endotracheal tube and ProSeal Laryngeal Mask Airway™ for laparoscopic gastric banding.
Anesthesiology (2012-05-23)
Michele Carron, Stefano Veronese, Walter Gomiero, Mirto Foletto, Donato Nitti, Carlo Ori, Ulderico Freo
ABSTRACT
The stress responses from tracheal intubation are potentially dangerous in patients with higher cardiovascular risk, such as obese patients. The primary outcome objective of this study was to test whether, in comparison with the endotracheal tube (ETT), the Proseal™ Laryngeal Mask Airway (PLMA™) (Laryngeal Mask Airway Company, Jersey, United Kingdom) reduces blood pressure and norepinephrine responses and the amounts of muscle relaxants needed in obese patients. We assessed hemodynamic and hormonal stress responses, ventilation, and postoperative recovery in 75 morbidly obese patients randomized to receive standardized anesthesia with either an ETT or the PLMA™ for laparoscopic gastric banding. In repeated-measures ANOVA, mean arterial blood pressure and plasma norepinephrine were significantly higher in the ETT group than in the PLMA™ group. In individual pairwise comparisons, blood pressure rose higher in ETT than PLMA™ patients after insertion and removal of airway devices, and after recovery. In ETT compared with PLMA™ patients, plasma norepinephrine was higher after induction of carboperitoneum (mean ± SD, 534 ± 198 and 368 ± 147 and pg/ml, P = 0.001), after airway device removal (578 ± 285 and 329 ± 128 pg/ml, P < 0.0001), and after recovery in postanesthesia care unit (380 ± 167 and 262 ± 95 and pg/ml, P = 0.003). Compared with use of the ETT, the PLMA™ reduced cisatracurium requirement, oxygen desaturation, and time to discharge from both the postanesthesia care unit and the hospital. PLMA™ reduces stress responses and postoperative complaints after laparoscopic gastric banding.
Product Description
Atracurium for peak identification, European Pharmacopoeia (EP) Reference Standard
Atracurium besylate, European Pharmacopoeia (EP) Reference Standard
Atracurium for impurity F identification, European Pharmacopoeia (EP) Reference Standard