MilliporeSigma
  • A retrospective case-control study comparing hysteroscopic resection versus hormonal modulation in treating menstrual disorders due to isthmocele.
A retrospective case-control study comparing hysteroscopic resection versus hormonal modulation in treating menstrual disorders due to isthmocele.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology (2011-01-06)
Pasquale Florio, Giampiero Gubbini, Elena Marra, Daniela Dores, Daniela Nascetti, Luca Bruni, Raffaele Battista, Irene Moncini, Marco Filippeschi, Felice Petraglia
ABSTRACT
In a retrospective case-control study, we compared the effectiveness of hysteroscopic correction and hormonal treatment to improve symptoms [postmestrual abnormal uterine bleeding (PAUB), pelvic pain localized in suprapubic site] associated with isthmocele. Women (n = 39; mean age ± SD, 35 ± 4.1 years) were subdivided in Group A [patients (n = 19) subjected to hysteroscopic surgery (isthmoplasty)] and, Group B [women (n = 20) undergoing hormonal treatment consisting of one oral tablet containing 0.075  mg of Gestodene and 0.030 mg of Ethynylestradiol for 21 days, followed by 7 days of suspension]. Resolution and/or improvement of menstrual disorders; patients degree of satisfaction with the treatment were measured 3 months later, by office hysteroscopy (Grop A) or phone call. PAUB and pelvic pain resolution was achieved in all patients: Group A had significant lower numbers of days of menstrual bleeding (P < 0.001), prevalence of pelvic pain in the suprapubic area (P = 0.04) and, higher degree of satisfaction (P < 0.001) compared to Group B. In conclusion, resectoscopic surgery is a valid way to treat patients with symptoms of prolonged postmenstrual uterine bleeding caused by isthmocele. Data from this study also indicate that resectoscopy may be the first choice because it is minimally invasive and yields good therapeutic results.
Sigma-Aldrich
Gestodene, ≥98% (HPLC)
Gestodene, European Pharmacopoeia (EP) Reference Standard