MilliporeSigma
  • Protective and damaging effects of platelets in acute cholestatic liver injury revealed by depletion and inhibition strategies.
Protective and damaging effects of platelets in acute cholestatic liver injury revealed by depletion and inhibition strategies.
Toxicological sciences : an official journal of the Society of Toxicology (2010-02-06)
Bradley P Sullivan, Ruipeng Wang, Ossama Tawfik, James P Luyendyk
ABSTRACT
Alpha-naphthylisothiocyanate (ANIT) causes cholestatic hepatitis characterized by intrahepatic bile duct epithelial cell injury and periportal hepatocellular necrosis. The progression of ANIT-induced hepatocyte injury is reported to involve extrahepatic cells including platelets. We showed recently that the procoagulant protein tissue factor (TF) is essential for ANIT-induced coagulation and contributes to ANIT-induced liver necrosis. Platelets have been shown to express TF and can contribute to coagulation cascade activation. To this end, we tested the hypothesis that platelet-dependent coagulation contributes to ANIT-induced liver injury. In ANIT (60 mg/kg)-treated mice, activation of the coagulation cascade occurred prior to a decrease of platelets in the blood. Immunostaining for glycoprotein IIb (CD41) revealed platelet accumulation along the borders of necrotic foci in livers of ANIT-treated mice. Antibody-mediated platelet depletion did not affect coagulation but markedly affected liver histopathology in ANIT-treated mice. Platelet depletion induced marked pooling of blood within necrotic lesions consistent with parenchymal-type peliosis as early as 24 h after ANIT treatment. In contrast, treatment with the P2Y(12) inhibitor clopidogrel significantly reduced ANIT-induced hepatocyte necrosis and serum alanine aminotransferase activity but did not exaggerate bleeding into necrotic foci. Clopidogrel also reduced hepatic neutrophil accumulation but did not affect induction of Intercellular adhesion molecule-1 or chemokine CxC motif ligand-1 messenger RNA expression in liver. The data indicate that ANIT-induced coagulation is platelet independent and that platelets contribute to ANIT-induced hepatocyte necrosis by promoting neutrophil accumulation. In contrast, severe thrombocytopenia induces parenchymal-type peliosis in the livers of ANIT-treated mice, a rare hepatic lesion associated with pooling of blood in the liver.
Sigma-Aldrich
1-Naphthyl isothiocyanate, 95%