MilliporeSigma
  • Design, synthesis, and bioactivity study of novel benzoylpyridazyl ureas.
Design, synthesis, and bioactivity study of novel benzoylpyridazyl ureas.
Journal of agricultural and food chemistry (2009-07-16)
Ranfeng Sun, Yonglin Zhang, Fuchun Bi, Qingmin Wang
ABSTRACT
A series of novel benzoylpyridazyl ureas were designed and synthesized from maleic anhydride and hydrazine monohydrate. These benzoylureas were identified by (1)H NMR spectroscopy and element analysis. The bioactivities of the new compounds were evaluated. These compounds exhibited larvicidal activities against oriental armyworm, and in particular, compound 13 displayed comparable activity to the commercial insecticide Hexaflumuron. Most of these compounds also had some larvicidal activities against mosquito. Interestingly, some compounds showed good plant growth regulatory activities.
Product Description
Supelco
Hexaflumuron, PESTANAL®, analytical standard