MilliporeSigma
  • NMR spectroscopy and molecular modelling studies of nitrosylcobalamin: further evidence that the deprotonated, base-off form is important for nitrosylcobalamin in solution.
NMR spectroscopy and molecular modelling studies of nitrosylcobalamin: further evidence that the deprotonated, base-off form is important for nitrosylcobalamin in solution.
Dalton transactions (Cambridge, England : 2003) (2009-01-06)
Hanaa A Hassanin, Luciana Hannibal, Donald W Jacobsen, Kenneth L Brown, Helder M Marques, Nicola E Brasch
ABSTRACT
The structure of nitrosylcobalamin (NOCbl) in solution has been studied by NMR spectroscopy and the 1H and 13C NMR spectra have been assigned. 13C and 31P NMR chemical shifts, the UV-vis spectrum of NOCbl and the observed pKbase-off value of approximately 5.1 for NOCbl provide evidence that a significant fraction of NOCbl is present in the base-off, 5,6-dimethylbenzimidazole (DMB) deprotonated, form in solution. NOE-restrained molecular mechanics modelling of base-on NOCbl gave annealed structures with minor conformational differences in the flexible side chains and the nucleotide loop position compared with the X-ray structure. A molecular dynamics simulation at 300 K showed that DMB remains in close proximity to the alpha face of the corrin in the base-off form of NOCbl. Simulated annealing calculations produced two major conformations of base-off NOCbl. In the first, the DMB is perpendicular to the corrin and its B3 nitrogen is about 3.1 A away from and pointing directly at the metal ion; in the second the DMB is parallel to and tucked beneath the D ring of the corrin.
Sigma-Aldrich
5,6-Dimethylbenzimidazole, ≥99%