MilliporeSigma
  • Resistance-induced changes in triclabendazole transport in Fasciola hepatica: ivermectin reversal effect.
Resistance-induced changes in triclabendazole transport in Fasciola hepatica: ivermectin reversal effect.
The Journal of parasitology (2007-02-20)
L Mottier, L Alvarez, I Fairweather, C Lanusse
ABSTRACT
Triclabendazole (TCBZ) and albendazole (ABZ) are flukicidal benzimidazole compounds extensively used in veterinary medicine. Although TCBZ has excellent activity against mature and immature stages of the liver fluke, Fasciola hepatica, ABZ action is restricted to flukes older than 12 wk. The intensive use of TCBZ has resulted in the development of resistance. To gain insight into the mechanisms of resistance to TCBZ, the ex vivo diffusion of TCBZ, TCBZ sulfoxide (TCBZSO, the active metabolite of TCBZ), and ABZ into TCBZ-susceptible and -resistant adult flukes was compared. TCBZ-susceptible (Cullompton) and -resistant (Sligo) flukes were incubated in Krebs-Ringer Tris buffer with either TCBZ, TCBZSO, or ABZ (5 nmol/ ml) for 90 min. Drug/metabolite concentrations were quantified by high-performance liquid chromatography. All the assayed molecules penetrated through the tegument of both susceptible and resistant flukes. However, significantly lower concentrations of TCBZ and TCBZSO were recovered within the TCBZ-resistant flukes. In contrast, ABZ entrance into the susceptible and resistant flukes was equivalent. The influx/efflux balance for TCBZ, TCBZSO, and ABZ in susceptible and resistant flukes in the presence or absence of a substrate (ivermectin) of the drug transporter P-glycoprotein was assessed. The ivermectin-induced modulation of P-glycoprotein activity decreased TCBZ efflux from the resistant flukes. Higher concentrations of TCBZ and TCBZSO were recovered from the resistant liver flukes in the presence of ivermectin. Thus, an altered influx/efflux mechanism may account for the development of resistance to TCBZ in F. hepatica.
Product Description
Supelco
Triclabendazole sulfoxide, VETRANAL®, analytical standard