MilliporeSigma
  • Effect of temperature on subsite map of Bacillus licheniformis alpha-amylase.
Effect of temperature on subsite map of Bacillus licheniformis alpha-amylase.
Acta biologica Hungarica (2006-10-20)
Lili Kandra, Judit Remenyik, Gyöngyi Gyémánt, A Lipták
ABSTRACT
To elucidate how temperature effects subsite mapping of a thermostable alpha-amylase from Bacillus licheniformis (BLA), a comparative study was performed by using 2-chloro-4-nitrophenyl (CNP) beta-maltooligosides with degree of polymerisation (DP) 4-10 as model substrates. Action patterns, cleavage frequencies and subsite binding energies were determined at 50 degrees C, 80 degrees C and 100 degrees C. Subsite map at 80 degrees C indicates more favourable bindings compared to the hydrolysis at 50 degrees C. Hydrolysis at 100 degrees C resulted in a clear shift in the product pattern and suggests significant differences in the active site architecture. Two preferred cleavage modes were seen for all substrates in which subsite (+2) and (+3) were dominant, but CNP-G1 was never formed. In the preferred binding mode of shorter oligomers, CNP-G2 serves as the leaving group (79%, 50%, 59% and 62% from CNP-G4, CNP-G5, CNP-G6 and CNP-G7, respectively), while CNP-G3 is the dominant hydrolysis product from CNP-G8, CNP-G9, and CNP-Gl0 (62%, 68% and 64%, respectively). The high binding energy value (-17.5 kJ/mol) found at subsite (+2) is consistent with the significant formation of CNP-G2. Subsite mapping at 80 degrees C and 100 degrees C confirms that there are no further binding sites despite the presence of longer products.
Sigma-Aldrich
4-Chloro-2-nitrophenol, ≥97.0%