MilliporeSigma
  • A novel route to dipeptides via noncondensation of amino acids: 2-aminoperfluoropropene as a synthon for trifluoroalanine dipeptides.
A novel route to dipeptides via noncondensation of amino acids: 2-aminoperfluoropropene as a synthon for trifluoroalanine dipeptides.
Organic letters (2006-02-24)
Yong Guo, Kana Fujiwara, Kenji Uneyama
ABSTRACT
2-Aminoperfluoropropene has been prepared by the Mg-promoted defluorinative N-silylation of N-p-methoxyphenyl hexafluoroacetone imine and has been employed as a synthon of trifluoroalanine for the preparation of trifluoroalanine dipeptides.
Sigma-Aldrich
3,3,3-Trifluoro-DL-alanine, 98%