MilliporeSigma
Parallel synthesis and purification using anthracene-tagged substrates.
Organic letters (2000-11-18)
X Wang, J J Parlow, J A Porco
ABSTRACT
[reaction: see text] A new strategy for the synthesis and purification of synthetic intermediates is described using anthracene-tagged ester substrates in conjunction with an N-benzylmaleimide resin. Anthracene chemical tags permit use of standard solution-phase reaction conditions and reaction-monitoring techniques.
Sigma-Aldrich
N-Benzylmaleimide, 99%