MilliporeSigma
  • Synthesis and antitrypanosomal evaluation of E-isomers of 5-nitro-2-furaldehyde and 5-nitrothiophene-2-carboxaldehyde semicarbazone derivatives. structure-activity relationships.
Synthesis and antitrypanosomal evaluation of E-isomers of 5-nitro-2-furaldehyde and 5-nitrothiophene-2-carboxaldehyde semicarbazone derivatives. structure-activity relationships.
European journal of medicinal chemistry (2000-04-29)
H Cerecetto, R Di Maio, M González, M Risso, G Sagrera, G Seoane, A Denicola, G Peluffo, C Quijano, A O Stoppani, M Paulino, C Olea-Azar, M A Basombrío
ABSTRACT
Several novel semicarbazone derivatives were prepared from 5-nitro-2-furaldehyde or 5-nitrothiophene-2-carboxaldehyde and semicarbazides bearing a spermidine-mimetic moiety. All derivatives presented the E-configuration, as determined by NMR-NOE experiments. These compounds were tested in vitro as potential antitrypanosomal agents, and some of them, together with the parent compounds, 5-nitro-2-furaldehyde and 5-nitrothiophene-2-carboxaldehyde semicarbazone derivatives, were also evaluated in vivo using infected mice. Structure-activity relationship studies were carried out using voltammetric response and lipophilic-hydrophilic balance as parameters. Two of the compounds (1 and 3) displayed the highest in vivo activity. A correlation was found between lipophilic-hydrophilic properties and trypanocidal activity, high R(M) values being associated with low in vivo effects.
Sigma-Aldrich
5-Nitro-2-furaldehyde, 99%