MilliporeSigma
  • High-performance liquid chromatographic determination and pharmacokinetic study of apigenin-7-O-β-D-glucoside in rat plasma after intravenous administration.
High-performance liquid chromatographic determination and pharmacokinetic study of apigenin-7-O-β-D-glucoside in rat plasma after intravenous administration.
Archives of pharmacal research (2011-06-10)
Zaixing Chen, Xixiang Ying, Shu Meng, Xu Zhu, Hong Jiang, Qishen Cao, Xuying Li, Fanhao Meng
ABSTRACT
The present study was designed to validate an analytical method based on HPLC for the quantitative measurement of the Apigenin-7-O-β-D-glucoside (AGL) in rat plasma after intravenous administration. HPLC analysis was done using a C18 column, UV detection at 335 nm, and a mobile phase of methanol:0.2% phosphoric acid (1:1, v/v). Good linearity was observed over the range of 0.06-7.20 μg/mL with a lower limit of quantication of 0.06 μg/mL. The intra-and inter-day precision values were below 9.97%. No interference peaks or matrix effects were observed. The method was fully validated and successfully applied to a pharmacokinetic study of AGL.
Product Description
Apigenin 7-glucoside, primary reference standard
Supelco
Apigenin 7-glucoside, analytical standard