MilliporeSigma
[Study on the flavonoids constituents of Trachelospermum jasminoides].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-06-04)
Xing-Qi Tan, Liang-Jun Guo, Hai-Sheng Chen, Lin-Song Wu, Fei-Fei Kong
ABSTRACT
To study the flavonoids constituents of Trachelospemum jasminoides. The compounds were separated and purified by column chromatography with silica gel, and identified by IR, MS, NMR and 2D-NMR. Six flavonoids were identified as apigenin (I), apigenin 7-O-beta-glucoside (II), apigenin 7-O-beta-neospheroside (III), luteoloside (IV), narngin (V) 6,8-di-C-glucopyanosylapigenin (VI), respectively. Compounds V and VI are isolated from this plant for the first time.
Product Description
Apigenin 7-glucoside, primary reference standard
Supelco
Apigenin 7-glucoside, analytical standard