MilliporeSigma
  • Purification, characterization, and crystallization of an N-hydroxyarylamine O-acetyltransferase from Salmonella typhimurium.
Purification, characterization, and crystallization of an N-hydroxyarylamine O-acetyltransferase from Salmonella typhimurium.
Protein expression and purification (1998-05-16)
J C Sinclair, R Delgoda, M E Noble, S Jarmin, N K Goh, E Sim
ABSTRACT
The N-hydroxyarylamine O-acetyltransferase from Salmonella typhimurium has been expressed as a histidine-tagged fusion protein in Escherichia coli and purified to apparent homogeneity using single-step immobilized metal ion chromatography. Sufficient quantities of the purified protein have been obtained to allow its characterization by physical methods including dynamic light scattering and electrospray mass spectrometry. The substrate specificity and temperature sensitivity of the enzymatic activity have also been assessed. The enzyme has been crystallized from sodium, potassium tartrate and X-ray diffraction data have been obtained to allow the identification of an orthorhombic unit cell, point group P21212, with dimensions a = 137 A, b = 223 A, and c = 105 A. These crystals will provide a route to a crystallographic determination of the structure of the protein.
Sigma-Aldrich
4-Iodoaniline, 98%