MilliporeSigma
  • Rutoxyl [rutin/4-acetamide-1-hydroxy-2,2,6,6-tetramethylpiperidinium] is a new member of the class of semi-natural products of high pharmacological potency.
Rutoxyl [rutin/4-acetamide-1-hydroxy-2,2,6,6-tetramethylpiperidinium] is a new member of the class of semi-natural products of high pharmacological potency.
Biochemistry and molecular biology international (1997-09-26)
D Metodiewa, J Skolimowski, A Kochman, S Karolczak
ABSTRACT
A novel complex, Rutoxyl [rutin/4-acetamide-1-hydroxy-2,2,6,6-tetramethylpiperidinium] was synthesized and its structure and anticancer activity were investigated. The results reported here are consistent with our idea, that the formation of such a complex of two biologically active molecules: polyphenolic flavonoid antioxidant (Rutin) and nitroxylamine of nitroxide antioxidant (Tempace), stabilized by hydrogen bond(s) can result in a supra-additive properties.
Sigma-Aldrich
4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, purum, ≥98.0% (HPLC)
Sigma-Aldrich
4-Acetamido-TEMPO, free radical, 97%