MilliporeSigma
  • Photoaffinity casting of a coumarin flag for rapid identification of ligand-binding sites within protein.
Photoaffinity casting of a coumarin flag for rapid identification of ligand-binding sites within protein.
Chemical communications (Cambridge, England) (2013-01-26)
Shota Morimoto, Takenori Tomohiro, Nobuyuki Maruyama, Yasumaru Hatanaka
ABSTRACT
A photo-switchable fluorescent flagging approach has been developed to identify photoaffinity-labeled peptides in target protein. Upon photochemical release of the ligand, the protein was newly modified with a coumarin in place of the previously attached biotin. It allowed us to simplify complex identification processes for labeled sites.
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Coumarin, primary reference standard
Supelco
Coumarin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland