MilliporeSigma
  • Reduced-dose craniospinal radiotherapy followed by tandem high-dose chemotherapy and autologous stem cell transplantation in patients with high-risk medulloblastoma.
Reduced-dose craniospinal radiotherapy followed by tandem high-dose chemotherapy and autologous stem cell transplantation in patients with high-risk medulloblastoma.
Neuro-oncology (2012-12-22)
Ki Woong Sung, Do Hoon Lim, Meong Hi Son, Soo Hyun Lee, Keon Hee Yoo, Hong Hoe Koo, Ji Hye Kim, Yeon-Lim Suh, Yoo Sook Joung, Hyung Jin Shin
ABSTRACT
We assessed the feasibility and effectiveness of reduced-dose craniospinal (CS) radiotherapy (RT) followed by tandem high-dose chemotherapy and autologous stem cell transplantation (HDCT/autoSCT) in reducing late adverse effects without jeopardizing survival among children with high-risk medulloblastoma (MB). From October 2005 through September 2010, twenty consecutive children aged >3 years with high-risk MB (presence of metastasis and/or postoperative residual tumor >1.5 cm(2)) were assigned to receive 2 cycles of pre-RT chemotherapy, CSRT (23.4 or 30.6 Gy) combined with local RT to the primary site (total 54.0 Gy), and 4 cycles of post-RT chemotherapy followed by tandem HDCT/autoSCT. Carboplatin-thiotepa-etoposide and cyclophosphamide-melphalan regimens were used for the first and second HDCT, respectively. Of 20 patients with high-risk MB, 17 had metastatic disease and 3 had a postoperative residual tumor >1.5 cm(2) without metastasis. The tumor relapsed/progressed in 4 patients, and 2 patients died of toxicities during the second HDCT/autoSCT. Therefore, 14 patients remained event-free at a median follow-up of 46 months (range, 23-82) from diagnosis. The probability of 5-year event-free survival was 70.0% ± 10.3% for all patients and 70.6% ± 11.1% for patients with metastases. Late adverse effects evaluated at a median of 36 months (range, 12-68) after tandem HDCT/autoSCT were acceptable. In children with high-risk MB, CSRT dose might be reduced when accompanied by tandem HDCT/autoSCT without jeopardizing survival. However, longer follow-up is needed to evaluate whether the benefits of reduced-dose CSRT outweigh the long-term risks of tandem HDCT/autoSCT.
Sigma-Aldrich
Thio-TEPA