MilliporeSigma
  • Hispidin analogue davallialactone attenuates carbon tetrachloride-induced hepatotoxicity in mice.
Hispidin analogue davallialactone attenuates carbon tetrachloride-induced hepatotoxicity in mice.
Journal of natural products (2012-10-03)
Prabodh Risal, Pyoung Han Hwang, Bong Sik Yun, Ho-Keun Yi, Baik Hwan Cho, Kyu Yun Jang, Yeon Jun Jeong
ABSTRACT
In this study the protective effects of davallialactone (1), isolated from Inonotus xeranticus, have been examined against carbon tetrachloride (CCl₄-induced acute liver injury. Mice received subcutaneous injection of 1 (2.5, 5, and 10 mg/kg) for three days before CCl₄ injection (1 mg/kg). Protection from liver injury by 1 was confirmed by the observation of decreased serum transaminases and diminished necrosis of liver tissue. Reduced hepatic injury was very similar to that observed with silymarin, a known hepatoprotective drug used in this work for comparison. The groups treated with 1 had reduced reactive oxygen species (ROS), reduced serum malonyldialdehyde levels, and increased levels of liver Cu/Zn superoxide dismutase, as compared to the CCl₄ control group. The expression of heme oxygenase-1 in the liver tissue was increased and the activity of liver cytochrome P4502E1 was restored in the mice treated with 1. In addition, levels of serum tumor necrosis factor-alpha (TNF-α), inducible NO synthase (iNOS), and cyclooxygenase-2 (COX-2), numbers of macrophage, and cleaved caspase-3-positive hepatocytes were reduced in the groups treated with 1. These findings suggest that davallialactone has protective effects against CCl₄-induced acute liver injury, and this protection is likely due to the suppression of ROS-induced lipid peroxidation and inflammatory response.
Sigma-Aldrich
Hispidin, solid, ≥98% (HPLC)