MilliporeSigma
  • Genome sequence of the moderately halotolerant, arsenite-oxidizing bacterium Pseudomonas stutzeri TS44.
Genome sequence of the moderately halotolerant, arsenite-oxidizing bacterium Pseudomonas stutzeri TS44.
Journal of bacteriology (2012-07-31)
Xiangyang Li, Jing Gong, Yao Hu, Lin Cai, Laurel Johnstone, Gregor Grass, Christopher Rensing, Gejiao Wang
ABSTRACT
We present the draft genome sequence of Pseudomonas stutzeri TS44, a moderately halotolerant, arsenite-oxidizing bacterium isolated from arsenic-contaminated soil. The genome contains genes for arsenite oxidation, arsenic resistance, and ectoine/hydroxyectoine biosynthesis. The genome information will be useful for exploring adaptation of P. stutzeri TS44 to an arsenic-contaminated environment.
Sigma-Aldrich
Ectoine, osmoprotectant
Sigma-Aldrich
Ectoine, ≥95.0% (HPLC)