MilliporeSigma
  • Delay of flower senescence by bacterial endophytes expressing 1-aminocyclopropane-1-carboxylate deaminase.
Delay of flower senescence by bacterial endophytes expressing 1-aminocyclopropane-1-carboxylate deaminase.
Journal of applied microbiology (2012-07-24)
S Ali, T C Charles, B R Glick
ABSTRACT
The ability of 1-aminocyclopropane-1-carboxylate (ACC) deaminase-containing plant growth-promoting bacterial (PGPB) endophytes Pseudomonas fluorescens YsS6 and Pseudomonas migulae 8R6, their ACC deaminase minus mutants and the rhizospheric plant growth-promoting bacterium Pseudomonas putida UW4 to delay the senescence of mini carnation cut flowers was assessed. Fresh cut flowers were incubated with either a bacterial cell suspension, the ethylene precursor ACC, the ethylene inhibitor l-α-(aminoethoxyvinyl)-glycine or 0·85% NaCl at room temperature for 11 days. Levels of flower senescence were recorded every other day. To verify the presence of endophytes inside the plant tissues, scanning electron microscopy was performed. Among all treatments, flowers treated with wild-type ACC deaminase-containing endophytic strains exhibited the most significant delay in flower senescence, while flowers treated with the ACC deaminase minus mutants senesced at a rate similar to the control. Flowers treated with Ps. putida UW4 senesced more rapidly than untreated control flowers. The only difference between wild-type and mutant bacterial endophytes was ACC deaminase activity so that it may be concluded that this enzyme is directly responsible for the significant delay in flower senescence. Despite containing ACC deaminase activity, Ps. putida UW4 is not taken up by the cut flowers and therefore has no effect on prolonging their shelf life. The world-wide cut flower industry currently uses expensive and potentially environmentally dangerous chemical inhibitors of ethylene to prolong the shelf life of cut flowers. The use of PGPB endophytes with ACC deaminase activity has the potential to replace the chemicals that are currently used by the cut flower industry.
Sigma-Aldrich
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride, ≥93% (AT)
Supelco
Aminoethoxyvinyl glycine hydrochloride, PESTANAL®, analytical standard