MilliporeSigma
  • Effect of sumatriptan on gastric emptying: a crossover study using the BreathID system.
Effect of sumatriptan on gastric emptying: a crossover study using the BreathID system.
World journal of gastroenterology (2012-07-19)
Yasunari Sakamoto, Yusuke Sekino, Eiji Yamada, Takuma Higurashi, Hidenori Ohkubo, Eiji Sakai, Hiroki Endo, Hiroshi Iida, Takashi Nonaka, Koji Fujita, Masato Yoneda, Tomoko Koide, Hirokazu Takahashi, Ayumu Goto, Yasunobu Abe, Eiji Gotoh, Shin Maeda, Atsushi Nakajima, Masahiko Inamori
ABSTRACT
To determine the effect of oral sumatriptan on gastric emptying using a continuous ¹³C breath test (BreathID system). Ten healthy male volunteers participated in this randomized, 2-way crossover study. The subjects fasted overnight and were randomly assigned to receive a test meal (200 kcal/200 mL) 30 min after pre-medication with sumatriptan 50 mg (sumatriptan condition), or the test meal alone (control condition). Gastric emptying was monitored for 4 h after administration of the test meal by the ¹³C-acetic acid breath test performed continually using the BreathID system. Then, using Oridion Research Software (β version), the time taken for emptying of 50% of the labeled meal (T(1/2)) similar to the scintigraphy lag time for 10% emptying of the labeled meal (T(lag)), the gastric emptying coefficient (GEC), and the regression-estimated constants (β and κ) were calculated. The statistical significance of any differences in the parameters were analyzed using Wilcoxon's signed-rank test. In the sumatriptan condition, significant differences compared with the control condition were found in T(1/2) [median 131.84 min (range, 103.13-168.70) vs 120.27 min (89.61-138.25); P = 0.0016], T(lag) [median 80.085 min (59.23-125.89) vs 61.11 min (39.86-87.05); P = 0.0125], and β [median 2.3374 (1.6407-3.8209) vs 2.0847 (1.4755-2.9269); P = 0.0284]. There were no significant differences in the GEC or κ between the 2 conditions. This study showed that oral sumatriptan significantly delayed gastric emptying of a liquid meal.
Product Description
Sumatriptan for system suitability, European Pharmacopoeia (EP) Reference Standard
Sumatriptan succinate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sumatriptan succinate, ≥98% (HPLC), solid