MilliporeSigma
  • Receptor-targeting phthalocyanine photosensitizer for improving antitumor photocytotoxicity.
Receptor-targeting phthalocyanine photosensitizer for improving antitumor photocytotoxicity.
PloS one (2012-06-14)
Peng Xu, Jincan Chen, Zhuo Chen, Shanyong Zhou, Ping Hu, Xueyuan Chen, Mingdong Huang
ABSTRACT
Photodynamic therapy (PDT) is a promising therapeutic modality which uses a photosensitizer to capture visible light resulting in phototoxicity in the irradiated region. PDT has been used in a number of pathological indications, including tumor. A key desirable feature of the photosensitizer is the high phototoxicity on tumor cells but not on normal cells. In this study, we conjugate a gonadotropin-releasing hormone (GnRH) to a photosensitizer, Zinc phthalocyanine (ZnPc), in order to enhance its specificity to breast cancer, which over-expresses GnRH receptor. ZnPc has unique advantages over other photosensitizers, but is difficult to derivatize and purify as a single isomer. We previously developed a straight-forward way to synthesize mono-substituted β-carboxy-phthalocyanine zinc (ZnPc-COOH). Photophysical and photochemical parameters of this ZnPc-GnRH conjugate including fluorescence quantum yield (Ф(f)), fluorescence decay time (τ(s)) and singlet oxygen quantum yield (Ф(Δ)) were evaluated and found comparable with that of ZnPc, indicating that addition of a GnRH peptide does not significantly alter the generation of singlet oxygen from ZnPc. Cellular uptakes and phototoxicities of this conjugate were tested and found significantly enhanced on human breast cancer cell lines overexpressing GnRH receptors (MDA-MB-231 and MCF-7 cells) compared to cells with low levels of GnRH receptors, such as human embryonic lung fibroblast (HELF) and human liver carcinoma (HepG2) cells. In addition, the cellular uptake of this conjugate toward MCF-7 cells were found clearly alleviated by a GnRH receptor blocker Cetrorelix, suggesting that the cellular uptake of this conjugate was GnRH receptor-mediated. Put together, these findings revealed that coupling ZnPc with GnRH analogue was an effective way to improve the selectivity of ZnPc towards tumors with over-expressed GnRH receptors.
Sigma-Aldrich
Zinc phthalocyanine, Dye content 97 %