MilliporeSigma
  • A myristicin-rich essential oil from Daucus sahariensis growing in Algeria.
A myristicin-rich essential oil from Daucus sahariensis growing in Algeria.
Natural product communications (2011-08-06)
Tahar Smaili, Amar Zellagui, Pier Luigi Cioni, Guido Flamini
ABSTRACT
The essential oils obtained by hydrodistillation from leaves and fruits of Daucus sahariensis Murb. were analyzed by GC/MS. The main constituents of the essential oil from the leaves were myristicin (34.3%), alpha-pinene (5.4%), cis-chrysanthenyl acetate (5.3%) and epi-alpha-bisabolol (4.8%), and those from the fruits myristicin (43.9%), alpha-pinene (13.1%), limonene (9.4%), and cis-chrysanthenyl acetate (7.4%). Myristicin, the main constituent of both essential oils, is generally absent in the oils from other Daucus species, permitting the hypothesis that this compound is a chemical marker of this Saharan species.
Product Description
Supelco
Myristicin, analytical standard
Sigma-Aldrich
Myristicin from parsley leaf oil, ≥85% (HPLC), oil