MilliporeSigma
  • Pharmacokinetics of colistin and colistimethate sodium after a single 80-mg intravenous dose of CMS in young healthy volunteers.
Pharmacokinetics of colistin and colistimethate sodium after a single 80-mg intravenous dose of CMS in young healthy volunteers.
Clinical pharmacology and therapeutics (2011-05-06)
W Couet, N Grégoire, P Gobin, P J Saulnier, D Frasca, S Marchand, O Mimoz
ABSTRACT
Colistin pharmacokinetics (PK) was investigated in young healthy volunteers after a 1-h infusion of 80 mg (1 million international units (MIU)) of the prodrug colistin methanesulfonate (CMS). Concentration levels of CMS and colistin were determined in plasma and urine using a new chromatographic assay and analyzed simultaneously with a population approach after correcting the urine-related data for postexcretion hydrolysis of CMS into colistin. CMS and colistin have low volumes of distribution (14.0 and 12.4 liters, respectively), consistent with distribution being restricted to extracellular fluid. CMS is mainly excreted unchanged in urine (70% on average), with a typical renal clearance estimated at 103 ml/min-close to the glomerular filtration rate. Colistin elimination is essentially extrarenal, given that its renal clearance is 1.9 ml/min, consistent with extensive reabsorption. Colistin elimination is not limited by the formation rate because its half-life (3 h) is longer than that of CMS. The values of these pharmacokinetic parameters will serve as reference points for future comparisons with patients' data.
Sigma-Aldrich
Colistin sodium methanesulfonate, ~11,500 U/mg
Sigma-Aldrich
Colistin sodium methanesulfonate, from Bacillus colistinus
Colistimethate sodium, European Pharmacopoeia (EP) Reference Standard