MilliporeSigma
  • N-D-Gluco-N-methylalkanamide compounds, a new class of non-ionic detergents for membrane biochemistry.
N-D-Gluco-N-methylalkanamide compounds, a new class of non-ionic detergents for membrane biochemistry.
The Biochemical journal (1982-11-01)
J E Hildreth
ABSTRACT
N-d-Gluco-N-methylalkanamide detergents have been synthesized. The detergents, which were produced in high yield and at low cost, compared favourably in biochemical studies with commonly used non-ionic detergents, including a chemically related n-alkyl glucoside. The ease of removal by dialysis, high solubilizing power and non-denaturing properties of this new class of detergents make them valuable reagents for membrane research.
Product Description
Avanti
MEGA-9, Avanti Research - A Croda Brand 850541P, powder
Avanti
MEGA-10, Avanti Research - A Croda Brand 850542P, powder
Avanti
MEGA-8, Avanti Research - A Croda Brand 850540P, powder