MilliporeSigma
  • Biotransformation of α-bromoacetophenones by the marine fungus Aspergillus sydowii.
Biotransformation of α-bromoacetophenones by the marine fungus Aspergillus sydowii.
Marine biotechnology (New York, N.Y.) (2009-11-27)
Lenilson Coutinho Rocha, Hercules Vicente Ferreira, Eli Fernando Pimenta, Roberto Gomes Souza Berlinck, Maria Olímpia Oliveira Rezende, Maria Diva Landgraf, Mirna Helena Regali Seleghim, Lara Durães Sette, André Luiz Meleiro Porto
ABSTRACT
The biotransformation reactions of α-bromoacetophenone (1), p-bromo-α-bromoacetophenone (2), and p-nitro-α-bromoacetophenone (3) by whole cells of the marine fungus Aspergillus sydowii Ce19 have been investigated. Fungal cells that had been grown in artificial sea water medium containing a high concentration of chloride ions (1.20 M) catalysed the biotransformation of 1 to 2-bromo-1-phenylethanol 4 (56%), together with the α-chlorohydrin 7 (9%), 1-phenylethan-1,2-diol 9 (26%), acetophenone 10 (4%) and phenylethanol 11 (5%) identified by GC-MS analysis. In addition, it was observed that the enzymatic reaction was accompanied by the spontaneous debromination of 1 to yield α-chloroacetophenone 5 (9%) and α-hydroxyacetophenone 6 (18%) identified by GC-FID analysis. When 2 and 3 were employed as substrates, various biotransformation products were detected but the formation of halohydrins was not observed. It is concluded that marine fungus A. sydowii Ce19 presents potential for the biotransformations of bromoacetophenone derivatives.
Sigma-Aldrich
2-Bromoacetophenone, 98%
Supelco
2-Bromoacetophenone, for GC derivatization, LiChropur, ≥99.0%