MilliporeSigma
  • Synthesis and cytotoxic evaluation of 6-(3-pyrazolylpropyl) derivatives of 1,4-naphthohydroquinone-1,4-diacetate.
Synthesis and cytotoxic evaluation of 6-(3-pyrazolylpropyl) derivatives of 1,4-naphthohydroquinone-1,4-diacetate.
Archiv der Pharmazie (2009-09-16)
Aurora Molinari, Alfonso Oliva, Claudia Ojeda, José M Miguel del Corral, M Angeles Castro, Carmen Cuevas, Arturo San Feliciano
ABSTRACT
Several new 6-(3-pyrazolylpropyl) derivatives of 1,4-naphthohydroquinone-1,4-diacetate (NHQ-DA) have been prepared by chemical modifications of the Diels-Alder adduct of alpha-myrcene and 1,4-benzoquinone. All these new compounds and precursors have been evaluated in vitro for their cytotoxicity against cultured human cancer cells of MB-231 breast-adeno carcinoma, A-549 lung carcinoma, and HT-29 colon carcinoma. GI(50) values ranged in and below the micromolar concentration level.
Sigma-Aldrich
1,4-Dihydroxynaphthalene, technical, ≥90% (HPLC)