MilliporeSigma
  • Synthesis of heteroaromatic tropeines and heterogeneous binding to glycine receptors.
Synthesis of heteroaromatic tropeines and heterogeneous binding to glycine receptors.
Bioorganic & medicinal chemistry (2009-09-04)
Gábor Maksay, Zoltán Vincze, Péter Nemes
ABSTRACT
Heteroaromatic carboxylic esters of (nor)tropine were synthesized. Tropine esters displaced [(3)H]strychnine binding to glycine receptors of rat spinal cord with low Hill slopes. Two-site displacement resulted in nanomolar IC(50,1) and micromolar IC(50,2) values, and IC(50,2)/IC(50,1) ratios up to 615 depending on the heteroaromatic rings and N-methyl substitution. Nortropeines displayed high affinity and low heterogeneity. IC(50,1) and IC(50,2) values of tropeines did not correlate suggesting different binding modes/sites. Glycine potentiated only the nanomolar displacement reflecting positive allosteric interactions and potentiation of ionophore function. Affinities of three (nor)tropeines were different for glycine receptors but identical for 5-HT(3) receptors.
Sigma-Aldrich
Tropine, ≥97.0% (NT)