MilliporeSigma
Long-term leaching test of organo-contaminated cement-clay pastes.
Journal of hazardous materials (2009-06-16)
L Zampori, P Gallo Stampino, G Dotelli
ABSTRACT
The aim of the present work is to investigate the effect of a prolonged leaching test (more than a year) on the microstructure of solidified cementitious wasteforms. A set of four different cement-based monoliths (Ap, Bp, Cp and Dp) was prepared, and for each series an uncontaminated sample was prepared as reference (A-D). An organoclay was added in all pastes as pre-sorbent material for the pollutant; a model liquid organic pollutant, 2-chloroaniline (2-CA), was added only in the contaminated ones and different types of admixtures, chosen among those typically employed in the concrete mix-design, were used. After the first 28 days of curing, all the monoliths, contaminated and uncontaminated, underwent a dynamic leach testing (DLT) for more than 1 year in deionized water.
Sigma-Aldrich
2-Chloroaniline, technical, ≥98.0% (GC)
Sigma-Aldrich
2-Chloroaniline, ≥99.5% (GC)